Learn More
Medchemexpress LLC Docetaxel-d5 trihydrate | 812.91 | C43H48D5NO14 | 500 UG
SDP
Catalog No. 5000284302
Encompass_Preferred
Docetaxel-d5 is a deuterium-labeled analogue of docetaxel intended for use as an internal standard or tracer in analytical workflows, particularly LC-MS/MS assays. It is provided as the trihydrate form with formula C43H48D5NO14 and molecular weight 812.91, packaged in microgram-scale quantities for calibration and quantitation.
  • Isotopically labeled (d5) analogue for use as an internal standard.
  • Suitable for LC-MS/MS and other mass spectrometry assays.
  • Provided as a trihydrate with defined molecular formula and molecular weight.
  • Supplied in microgram-scale quantities for assay calibration.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-84302 Supplier Medchemexpress LLC Supplier No. HYW653942500UG
Docetaxel-d5 is a deuterium-labeled analogue of docetaxel intended for use as an internal standard or tracer in analytical workflows, particularly LC-MS/MS assays. It is provided as the trihydrate form with formula C43H48D5NO14 and molecular weight 812.91, packaged in microgram-scale quantities for calibration and quantitation.
  • Isotopically labeled (d5) analogue for use as an internal standard.
  • Suitable for LC-MS/MS and other mass spectrometry assays.
  • Provided as a trihydrate with defined molecular formula and molecular weight.
  • Supplied in microgram-scale quantities for assay calibration.
  • Intended for research use only; not for human or veterinary use.