Learn More
Cayman Chemical Docetaxel-d9, Purity: ≥99% deuterated forms (d1-d9), CAS: 940867-25-4, 1 mg
SDP
Catalog No. 502964263
Encompass_Preferred
An internal standard for the quantification of docetaxel by GC- or LC-MS
Catalog No. 50-296-4263 Supplier Cayman Chemical Supplier No. 220941 MG
An internal standard for the quantification of docetaxel by GC- or LC-MS