Learn More
Cayman Chemical Docetaxel-d9 500ug
SDP
Catalog No. 502987064
Encompass_Preferred
An internal standard for the quantification of docetaxel by GC- or LC-MS
Catalog No. 50-298-7064 Supplier Cayman Chemical Supplier No. 22094500 UG
An internal standard for the quantification of docetaxel by GC- or LC-MS