Learn More
Medchemexpress LLC Docetaxel-d9 | 940867-25-4 | 98.65% | C43H44D9NO14 | 1 MG
SDP
Catalog No. 5000432730
Encompass_Preferred
Docetaxel-d9 is a deuterium-labeled version of Docetaxel, known to act as a microtubule depolymerization inhibitor. It influences gene expression, causes cell cycle arrest, and induces cell apoptosis, demonstrating anti-cancer activity. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Microtubule depolymerization inhibitor
  • Reduces bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M
  • Induces cell apoptosis
Catalog No. 50-004-32730 Supplier Medchemexpress LLC Supplier No. HYB0011S1MG
Docetaxel-d9 is a deuterium-labeled version of Docetaxel, known to act as a microtubule depolymerization inhibitor. It influences gene expression, causes cell cycle arrest, and induces cell apoptosis, demonstrating anti-cancer activity. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Microtubule depolymerization inhibitor
  • Reduces bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M
  • Induces cell apoptosis