Learn More
Medchemexpress LLC Docetaxel (standard) | 114977-28-5 | 99.9% | C43H53NO14 | 100 MG
SDP
Catalog No. 5000432954
Encompass_Preferred
Docetaxel (standard) is an analytical standard of Docetaxel, developed for research and analytical applications. It functions as a microtubule depolymerization inhibitor and attenuates the effects of bcl-2 and bcl-xL gene expression. This compound arrests the cell cycle at G2/M and induces cell apoptosis, demonstrating anti-cancer activity.
  • Acts as a microtubule depolymerization inhibitor
  • Attenuates bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M
  • Induces cell apoptosis
  • Utilized in qualitative, quantitative, and methodological research experiments (e.g., HPLC, GC, MS)
  • Appears as a white to off-white solid
Catalog No. 50-004-32954 Supplier Medchemexpress LLC Supplier No. HYB0011R100MG
Docetaxel (standard) is an analytical standard of Docetaxel, developed for research and analytical applications. It functions as a microtubule depolymerization inhibitor and attenuates the effects of bcl-2 and bcl-xL gene expression. This compound arrests the cell cycle at G2/M and induces cell apoptosis, demonstrating anti-cancer activity.
  • Acts as a microtubule depolymerization inhibitor
  • Attenuates bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M
  • Induces cell apoptosis
  • Utilized in qualitative, quantitative, and methodological research experiments (e.g., HPLC, GC, MS)
  • Appears as a white to off-white solid