Learn More
Medchemexpress LLC Docetaxel (standard) | 114977-28-5 | 99.9% | C43H53NO14 | 5 MG
SDP
Catalog No. 5000432201
Encompass_Preferred
Docetaxel (Standard) is the analytical standard of Docetaxel, intended for research and analytical applications. Docetaxel (RP-56976) acts as a microtubule depolymerization inhibitor with an IC50 of 0.2 μM. It attenuates the effects of bcl-2 and bcl-xL gene expression, arrests the cell cycle at G2/M, and induces cell apoptosis, demonstrating anti-cancer activity.
  • Serves as a grade of analytical standard, providing a reference standard for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS
Catalog No. 50-004-32201 Supplier Medchemexpress LLC Supplier No. HYB0011R5MG
Docetaxel (Standard) is the analytical standard of Docetaxel, intended for research and analytical applications. Docetaxel (RP-56976) acts as a microtubule depolymerization inhibitor with an IC50 of 0.2 μM. It attenuates the effects of bcl-2 and bcl-xL gene expression, arrests the cell cycle at G2/M, and induces cell apoptosis, demonstrating anti-cancer activity.
  • Serves as a grade of analytical standard, providing a reference standard for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS