Learn More
Medchemexpress LLC Docetaxel (Standard) | 114977-28-5 | 99.9% | C43H53NO14 | 50 MG
SDP
Catalog No. 5000432264
Encompass_Preferred
Docetaxel (Standard) is an analytical standard intended for research and analytical applications. It acts as a microtubule depolymerization inhibitor, reducing the effects of bcl-2 and bcl-xL gene expression, arresting the cell cycle at G2/M, and inducing cell apoptosis. This demonstrates anti-cancer activity and makes it suitable for various research experiments.
  • Analytical standard for research and analytical applications
  • Microtubule depolymerization inhibitor
  • Reduces effects of bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M and induces cell apoptosis
  • Demonstrates anti-cancer activity
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-32264 Supplier Medchemexpress LLC Supplier No. HYB0011R50MG
Docetaxel (Standard) is an analytical standard intended for research and analytical applications. It acts as a microtubule depolymerization inhibitor, reducing the effects of bcl-2 and bcl-xL gene expression, arresting the cell cycle at G2/M, and inducing cell apoptosis. This demonstrates anti-cancer activity and makes it suitable for various research experiments.
  • Analytical standard for research and analytical applications
  • Microtubule depolymerization inhibitor
  • Reduces effects of bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M and induces cell apoptosis
  • Demonstrates anti-cancer activity
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications