Learn More
Medchemexpress LLC Docetaxel Standard | 114977-28-5 | C43H53NO14 | 25MG
SDP
Catalog No. 5000357883
Encompass_Preferred
Docetaxel (RP-56976) is a microtubule depolymerization inhibitor with an IC50 of 0.2 μM. It attenuates the effects of bcl-2 and bcl-xL gene expression, arrests the cell cycle at G2/M, and leads to cell apoptosis. Docetaxel exhibits anti-cancer activity.
  • Microtubule depolymerization inhibitor
  • Attenuates bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M
  • Leads to cell apoptosis
  • Exhibits anti-cancer activity
Catalog No. 50-003-57883 Supplier Medchemexpress LLC Supplier No. 25MGHYB0011R
Docetaxel (RP-56976) is a microtubule depolymerization inhibitor with an IC50 of 0.2 μM. It attenuates the effects of bcl-2 and bcl-xL gene expression, arrests the cell cycle at G2/M, and leads to cell apoptosis. Docetaxel exhibits anti-cancer activity.
  • Microtubule depolymerization inhibitor
  • Attenuates bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M
  • Leads to cell apoptosis
  • Exhibits anti-cancer activity