Learn More
Selleck Chemical LLC Docetaxel Trihydrate
SDP
Catalog No. 5000003796
Encompass_Preferred
Docetaxel (RP56976 NSC 628503) an analog of paclitaxel is an inhibitor of depolymerisation of microtubules by binding to stabilized microtubules
Catalog No. 50-000-03796 Supplier Selleck Chemical LLC Supplier No. S778750MG
Docetaxel (RP56976 NSC 628503) an analog of paclitaxel is an inhibitor of depolymerisation of microtubules by binding to stabilized microtubules