Learn More
Cayman Chemical Docetaxel (hydrate), Purity: ≥98%, CAS: 700367-34-6, 50 mg
SDP
Catalog No. 502951839
Encompass_Preferred
A microtubule-stabilizing agent; promotes microtubule assembly in a cell-free assay at 20 µM; is cytotoxic to J774.2 cells (EC50 = 0.05 µM); inhibits tumor growth in several cancer models, including B16 murine melanoma and P388 murine leukemia models
Catalog No. 50-295-1839 Supplier Cayman Chemical Supplier No. 1163750 MG
A microtubule-stabilizing agent; promotes microtubule assembly in a cell-free assay at 20 µM; is cytotoxic to J774.2 cells (EC50 = 0.05 µM); inhibits tumor growth in several cancer models, including B16 murine melanoma and P388 murine leukemia models