Learn More
Aviva Systems Biology Corp DOCK2 Antibody - middle region (ARP52058_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52058_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DOCK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DOCK2. Alternate names: FLJ46592, KIAA0209ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60677