Learn More
Enzo Life Sciences Docosa-4Z,7Z,10Z,13Z,16Z,19Z-hexaenoic acid (100mg). CAS: 6217-54-5
SDP
Catalog No. 502010791
Encompass_Preferred
Polyunsaturated fatty acid. Purity: ≥98%. Solubility: Soluble in ethanol, hexane and methylene chloride. Long Term Storage: -20°C.
Catalog No. 50-201-0791 Supplier Enzo Life Sciences Supplier No. BMLFA0020100
Polyunsaturated fatty acid. Purity: ≥98%. Solubility: Soluble in ethanol, hexane and methylene chloride. Long Term Storage: -20°C.