Learn More
Cayman Chemical Docosaenoyl EthnolamIde 10mg
SDP
Catalog No. 502893844
Encompass_Preferred
A long-chain ethanolamine that occurs at relatively high levels in rat CSF and presumably acts as an endocannabinoid
Catalog No. 50-289-3844 Supplier Cayman Chemical Supplier No. 900174510 MG
A long-chain ethanolamine that occurs at relatively high levels in rat CSF and presumably acts as an endocannabinoid