Learn More
Cayman Chemical Docosaenoyl Ethanolamide, Purity: ≥98%, CAS: 18190-74-4, 50 mg
SDP
Catalog No. 502895065
Encompass_Preferred
A long-chain ethanolamine that occurs at relatively high levels in rat CSF and presumably acts as an endocannabinoid
Catalog No. 50-289-5065 Supplier Cayman Chemical Supplier No. 900174550 MG
A long-chain ethanolamine that occurs at relatively high levels in rat CSF and presumably acts as an endocannabinoid