Learn More
Cayman Chemical DocosahexaenoIc ACD 1 2 3 4
SDP
Catalog No. 502989738
Encompass_Preferred
An internal standard for the quantification of DHA by GC- or LC-MS
Catalog No. 50-298-9738 Supplier Cayman Chemical Supplier No. 31104250 UG
An internal standard for the quantification of DHA by GC- or LC-MS