Learn More
Cayman Chemical DocosahexaenoIc ACD Alkyn
SDP
Catalog No. 502967479
Encompass_Preferred
An ω-alkyne derivative of docosahexaenoic acid; can be joined with azide-containing molecules using click chemistry; can be used track fatty acid metabolism in cells via click chemistry linkage to fluorophores
Catalog No. 50-296-7479 Supplier Cayman Chemical Supplier No. 16689100 UG
An ω-alkyne derivative of docosahexaenoic acid; can be joined with azide-containing molecules using click chemistry; can be used track fatty acid metabolism in cells via click chemistry linkage to fluorophores