Learn More
Cayman Chemical Docosahexaenoic Acid ethyl ester, Purity: ≥98%, CAS: 81926-94-5, 500 mg
SDP
Catalog No. 502892309
Encompass_Preferred
An esterified form of DHA; increases plasma and erythrocyte membrane DHA levels in rats without altering arachidonic acid levels when administered in the diet to at a dose equivalent to 8 mg/kg of DHA; reduces the number of reference memory errors in an eight-arm radial maze in 100-week old rats at 300 mg/kg
Catalog No. 50-289-2309 Supplier Cayman Chemical Supplier No. 9090310500 MG
An esterified form of DHA; increases plasma and erythrocyte membrane DHA levels in rats without altering arachidonic acid levels when administered in the diet to at a dose equivalent to 8 mg/kg of DHA; reduces the number of reference memory errors in an eight-arm radial maze in 100-week old rats at 300 mg/kg