Learn More
Cayman Chemical DocosahexaenoIc ACD ethyl S
SDP
Catalog No. 502892338
Encompass_Preferred
An internal standard for the quantification of docosahexaenoic acid ethyl ester by GC- or LC-MS
Catalog No. 50-289-2338 Supplier Cayman Chemical Supplier No. 9001245500 UG
An internal standard for the quantification of docosahexaenoic acid ethyl ester by GC- or LC-MS