Learn More
Cayman Chemical Docosahexaenoic Acid ethyl ester-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 2692624-15-8, 1 mg
SDP
Catalog No. 502894302
Encompass_Preferred
An internal standard for the quantification of docosahexaenoic acid ethyl ester by GC- or LC-MS
Catalog No. 50-289-4302 Supplier Cayman Chemical Supplier No. 90012451 MG
An internal standard for the quantification of docosahexaenoic acid ethyl ester by GC- or LC-MS