Learn More
Cayman Chemical DocosahexaenoIc ACD ethyl50UG
SDP
Catalog No. 502892421
Encompass_Preferred
An internal standard for the quantification of docosahexaenoic acid ethyl ester by GC- or LC-MS
Catalog No. 50-289-2421 Supplier Cayman Chemical Supplier No. 900124550 UG
An internal standard for the quantification of docosahexaenoic acid ethyl ester by GC- or LC-MS