Learn More
Cayman Chemical DocosahexaenoIc ACDd5 methy
SDP
Catalog No. 502989294
Encompass_Preferred
An internal standard for the quantification of DHA methyl ester by GC- or LC-MS
Catalog No. 50-298-9294 Supplier Cayman Chemical Supplier No. 31125100 UG
An internal standard for the quantification of DHA methyl ester by GC- or LC-MS