Learn More
Cayman Chemical Docosahexaenoic Acid, Purity: ≥98%, CAS: 6217-54-5, 100 mg
SDP
Catalog No. 502892979
Encompass_Preferred
A long-chain ω-3 PUFA; comprises approximately 40% of total brain PUFAs and is abundant in grey matter and retinal membranes; typically represents 0.52-7.5% of human total plasma fatty acids; has roles in several physiological and pathological processes
Catalog No. 50-289-2979 Supplier Cayman Chemical Supplier No. 90310100 MG
A long-chain ω-3 PUFA; comprises approximately 40% of total brain PUFAs and is abundant in grey matter and retinal membranes; typically represents 0.52-7.5% of human total plasma fatty acids; has roles in several physiological and pathological processes