Learn More
Medchemexpress LLC Docosahexaenoic acid 250mg | 81926-94-5 | 250MG
SDP
Catalog No. 5000239789
Encompass_Preferred
Docosahexaenoic acid ethyl ester (Ethyl docosahexaenoate) is a 90% concentrated ethyl ester of docosahexaenoic acid manufactured from the microalgal oil Docosahexaenoic acid ethyl ester enhances 6-hydroxydopamine-induced neuronal damage by induction of lipid peroxidation in mouse striatum Docosahexaenoic acid (DHA) is a key component of the cell membrane and its peroxidation is inducible due to the double-bond chemical structure Docosahexaenoic acid has neuroprotective effects[1 [2
Catalog No. 50-002-39789 Supplier Medchemexpress LLC Supplier No. HY107343250MG
Docosahexaenoic acid ethyl ester (Ethyl docosahexaenoate) is a 90% concentrated ethyl ester of docosahexaenoic acid manufactured from the microalgal oil Docosahexaenoic acid ethyl ester enhances 6-hydroxydopamine-induced neuronal damage by induction of lipid peroxidation in mouse striatum Docosahexaenoic acid (DHA) is a key component of the cell membrane and its peroxidation is inducible due to the double-bond chemical structure Docosahexaenoic acid has neuroprotective effects[1 [2