Learn More
Medchemexpress LLC Docosahexaenoic acid | 6217-54-5 | 98.8% | 200 MG
SDP
Catalog No. 5000411604
Encompass_Preferred
Docosahexaenoic acid (DHA) is an omega-3 fatty acid abundantly present in the brain and retina, crucial for development and maintaining normal function. Obtained from fish oil and maternal milk, DHA plays a vital role in cognitive abilities and neurological health.
  • Essential for infant brain growth and functional development
  • Maintains normal adult brain function
  • Improves learning ability and memory
  • Required for neural cell proliferation, migration, differentiation, and synaptogenesis
  • Unique structure supports effective cell signaling
  • Potent RXR ligand, activating RXRα at low micromolar concentrations
  • Reduces reference memory errors
  • Increases DHA content and DHA/arachidonic acid ratio in brain regions
  • Exhibits neuroprotective actions in Parkinson's disease models
Catalog No. 50-004-11604 Supplier Medchemexpress LLC Supplier No. HYB2167200MG
Docosahexaenoic acid (DHA) is an omega-3 fatty acid abundantly present in the brain and retina, crucial for development and maintaining normal function. Obtained from fish oil and maternal milk, DHA plays a vital role in cognitive abilities and neurological health.
  • Essential for infant brain growth and functional development
  • Maintains normal adult brain function
  • Improves learning ability and memory
  • Required for neural cell proliferation, migration, differentiation, and synaptogenesis
  • Unique structure supports effective cell signaling
  • Potent RXR ligand, activating RXRα at low micromolar concentrations
  • Reduces reference memory errors
  • Increases DHA content and DHA/arachidonic acid ratio in brain regions
  • Exhibits neuroprotective actions in Parkinson's disease models