Learn More
BLD PHARMATECH CO LIMITED Docosahexaenoic Acid, CAS Number: 6217-54-5, 5g, Purity: 95%
Catalog No. NC3965487
Encompass_Preferred
Docosahexaenoic Acid, CAS Number: 6217-54-5, 5g, Purity: 95%
Catalog No. NC3965487 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD1216515G
Docosahexaenoic Acid, CAS Number: 6217-54-5, 5g, Purity: 95%