Learn More
Medchemexpress LLC Docosahexaenoic acid derivative | 92510-91-3 | 98.16% | 419.60 | 1 ML
SDP
Catalog No. 5000401738
Encompass_Preferred
F 16915, a docosahexaenoic acid (DHA) derivative, is a potent pro-agent of DHA that can prevent heart failure-induced atrial fibrillation.
  • Potent pro-agent of docosahexaenoic acid (DHA).
  • Can prevent heart failure-induced atrial fibrillation.
  • Reduces marked dilation of the left atrial (LA) cavity and the steep decline of left ventricular (LV) contractile function.
  • Significantly reduces infarct size and prevents left ventricular free wall thinning.
  • Reduces the de-phosphorylation of connexin43 from atria tissue.
Catalog No. 50-004-01738 Supplier Medchemexpress LLC Supplier No. HY1988610MM/1ML
F 16915, a docosahexaenoic acid (DHA) derivative, is a potent pro-agent of DHA that can prevent heart failure-induced atrial fibrillation.
  • Potent pro-agent of docosahexaenoic acid (DHA).
  • Can prevent heart failure-induced atrial fibrillation.
  • Reduces marked dilation of the left atrial (LA) cavity and the steep decline of left ventricular (LV) contractile function.
  • Significantly reduces infarct size and prevents left ventricular free wall thinning.
  • Reduces the de-phosphorylation of connexin43 from atria tissue.