Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Docosahexaenoic acid ethyl ester | 81926-94-5 | 96.2% | 356.54 g/mol | C24H36O2 | 100 MG
SDP

Catalog No. 5000239876
Encompass_Preferred

Docosahexaenoic acid ethyl ester is the ethyl-ester derivative of the omega-3 fatty acid DHA, provided as a research-grade material for studies of lipid metabolism, neurobiology, and formulation work where the esterified form is required.

  • High purity (96.2%).
  • Molecular formula C24H36O2.
  • Molecular weight 356.54 g/mol.
  • Supplied as 100 mg in a sealed vial.
  • Suitable for in vitro and formulation studies involving omega-3 fatty acids.

Catalog No. 50-002-39876 Supplier Medchemexpress LLC Supplier No. HY107343100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Docosahexaenoic acid ethyl ester is the ethyl-ester derivative of the omega-3 fatty acid DHA, provided as a research-grade material for studies of lipid metabolism, neurobiology, and formulation work where the esterified form is required.

  • High purity (96.2%).
  • Molecular formula C24H36O2.
  • Molecular weight 356.54 g/mol.
  • Supplied as 100 mg in a sealed vial.
  • Suitable for in vitro and formulation studies involving omega-3 fatty acids.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.