Learn More
Cayman Chemical Docosahexaenoic Acid methyl e
SDP
Catalog No. 502888783
Encompass_Preferred
An esterified form of DHA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils
Catalog No. 50-288-8783 Supplier Cayman Chemical Supplier No. 10006865500 MG
Add to Cart
Add to Cart
An esterified form of DHA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils