Learn More
Cayman Chemical Docosahexaenoic Acid methyl e
SDP
Catalog No. 502889453
Encompass_Preferred
An esterified form of DHA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils
Catalog No. 50-288-9453 Supplier Cayman Chemical Supplier No. 100068651 G
Add to Cart
Add to Cart
An esterified form of DHA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils