Learn More
Medchemexpress LLC Docosahexaenoic acid methyl ester | 2566-90-7 | MFCD00673450 | 95.0% | 342.51 g/mol | C23H34O2 | 500 MG
SDP
Catalog No. 5000248042
Encompass_Preferred
Docosahexaenoic acid methyl ester is a methylated analog of docosahexaenoic acid used in research to incorporate DHA into membrane phospholipids while resisting oxidation and hydrolysis. It is provided as a high-purity analytical standard with documented solubility and in vivo formulation guidance.
  • Methylated DHA analog that resists oxidation and hydrolysis.
  • Intercalates into membrane phospholipids for membrane studies.
  • High purity: 95.0%.
  • Soluble in DMSO and ethanol; in vivo formulation examples provided.
  • Available in multiple package sizes, including a 500 mg pack.
Catalog No. 50-002-48042 Supplier Medchemexpress LLC Supplier No. HY101541500MG
Docosahexaenoic acid methyl ester is a methylated analog of docosahexaenoic acid used in research to incorporate DHA into membrane phospholipids while resisting oxidation and hydrolysis. It is provided as a high-purity analytical standard with documented solubility and in vivo formulation guidance.
  • Methylated DHA analog that resists oxidation and hydrolysis.
  • Intercalates into membrane phospholipids for membrane studies.
  • High purity: 95.0%.
  • Soluble in DMSO and ethanol; in vivo formulation examples provided.
  • Available in multiple package sizes, including a 500 mg pack.