Learn More
Cayman Chemical Docosahexaenoyl Serinol, Purity: ≥95%, CAS: 1638355-62-0, 10 mg
SDP
Catalog No. 502911385
Encompass_Preferred
A derivative of 2-DG; inhibits the growth of A549 cells (IC50 = 0.48 µM); inhibits degranulation of RBL-2H3 mast cells (IC50 = 16 µM); suppresses the IgE-dependent PCA reaction by 17% in mice at 20 mg/animal
Catalog No. 50-291-1385 Supplier Cayman Chemical Supplier No. 3567810 MG
A derivative of 2-DG; inhibits the growth of A549 cells (IC50 = 0.48 µM); inhibits degranulation of RBL-2H3 mast cells (IC50 = 16 µM); suppresses the IgE-dependent PCA reaction by 17% in mice at 20 mg/animal