Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Docosanedioic acid | 505-56-6 | MFCD00002806 | 97.0% | 370.57 | C22H42O4 | 250 MG
SDP

Catalog No. 5000202261
Encompass_Preferred

Docosanedioic acid is a saturated dicarboxylic acid used as a non-cleavable ADC linker and as an alkyl chain-based PROTAC linker in the synthesis of antibody-drug conjugates and PROTACs. It is supplied as an off-white to light yellow solid with high purity for research use.

  • Chemical formula: C22H42O4.
  • Molecular weight: 370.57.
  • CAS number: 505-56-6.
  • Purity: 97.0%.
  • Appearance: solid, off-white to light yellow.
  • Storage: powder at -20°C for long-term storage, 4°C for shorter storage; in solvent store at -80°C for extended stability.
  • Packaging: available in 250 mg, 1 g, 5 g, and 10 g sizes.
  • Applications: non-cleavable ADC linker and alkyl chain-based PROTAC linker for synthetic workflows.

Catalog No. 50-002-02261 Supplier Medchemexpress LLC Supplier No. HYW034918250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Docosanedioic acid is a saturated dicarboxylic acid used as a non-cleavable ADC linker and as an alkyl chain-based PROTAC linker in the synthesis of antibody-drug conjugates and PROTACs. It is supplied as an off-white to light yellow solid with high purity for research use.

  • Chemical formula: C22H42O4.
  • Molecular weight: 370.57.
  • CAS number: 505-56-6.
  • Purity: 97.0%.
  • Appearance: solid, off-white to light yellow.
  • Storage: powder at -20°C for long-term storage, 4°C for shorter storage; in solvent store at -80°C for extended stability.
  • Packaging: available in 250 mg, 1 g, 5 g, and 10 g sizes.
  • Applications: non-cleavable ADC linker and alkyl chain-based PROTAC linker for synthetic workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.