Learn More
Medchemexpress LLC Docosanoic acid | 112-85-6 | 99.4% | 340.59 | 100 G
SDP
Catalog No. 5000451416
Encompass_Preferred
Docosanoic acid (Behenic acid) is a long-chain saturated fatty acid. It inhibits the double-stranded DNA (dsDNA) binding activity of the p53 DNA binding domain, with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans.
  • Effectively inhibits dsDNA binding activity of wild-type p53 DBD with 50% inhibition at 0.41 nmol.
  • Significantly reduces deposition of visceral fat and increases fatty acid excretion in feces in rats.
  • Low bioavailability leads to reduced absorption, contributing to a reduction in body weight and visceral fat deposition.
Catalog No. 50-004-51416 Supplier Medchemexpress LLC Supplier No. HYW013049100G
Add to Cart
Add to Cart
Docosanoic acid (Behenic acid) is a long-chain saturated fatty acid. It inhibits the double-stranded DNA (dsDNA) binding activity of the p53 DNA binding domain, with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans.
  • Effectively inhibits dsDNA binding activity of wild-type p53 DBD with 50% inhibition at 0.41 nmol.
  • Significantly reduces deposition of visceral fat and increases fatty acid excretion in feces in rats.
  • Low bioavailability leads to reduced absorption, contributing to a reduction in body weight and visceral fat deposition.