Learn More
Medchemexpress LLC Docosanoic acid | 112-85-6 | 99.4% | 340.59 | 5 G
SDP
Catalog No. 5000454068
Encompass_Preferred
Docosanoic acid (Behenic acid) is a long-chain saturated fatty acid. It inhibits the double-stranded DNA (dsDNA) binding activity of p53 DNA binding domain, with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans. This product is for research use only; not sold to patients.
  • Appearance: solid, off-white to light yellow
  • Initial source: endogenous metabolite
  • Structure classification: ketones, aldehydes, acids
  • Inhibits dsDNA binding activity of wild-type p53 DBD
  • May reduce deposition of visceral fat in rats
Catalog No. 50-004-54068 Supplier Medchemexpress LLC Supplier No. HYW0130495G
Add to Cart
Add to Cart
Docosanoic acid (Behenic acid) is a long-chain saturated fatty acid. It inhibits the double-stranded DNA (dsDNA) binding activity of p53 DNA binding domain, with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans. This product is for research use only; not sold to patients.
  • Appearance: solid, off-white to light yellow
  • Initial source: endogenous metabolite
  • Structure classification: ketones, aldehydes, acids
  • Inhibits dsDNA binding activity of wild-type p53 DBD
  • May reduce deposition of visceral fat in rats