Learn More
Medchemexpress LLC Docosanoic acid-d3 (Behenic acid-d3) | 1193721-67-3 | HY-W013049S3 | 99.8% | 343.60 | 25 MG
SDP
Catalog No. 5000452047
Encompass_Preferred
Docosanoic acid-d3 (Behenic acid-d3) is the deuterium-labeled Docosanoic acid. It is a long-chain saturated fatty acid that inhibits the double-stranded DNA (dsDNA) binding activity of p53 DNA binding domain. Docosanoic acid has low bioavailability and can increase cholesterol in humans.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-52047 Supplier Medchemexpress LLC Supplier No. 25MGHYW013049S3
Docosanoic acid-d3 (Behenic acid-d3) is the deuterium-labeled Docosanoic acid. It is a long-chain saturated fatty acid that inhibits the double-stranded DNA (dsDNA) binding activity of p53 DNA binding domain. Docosanoic acid has low bioavailability and can increase cholesterol in humans.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs