Learn More
Medchemexpress LLC Docosanoic acid-d4-1 | 1219804-98-4 | 99.0% | 344.61 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW013049S410MG
Encompass_Preferred
Docosanoic acid-d4-1 (Behenic acid-d4-1) is the deuterium labeled Docosanoic acid. It is a long-chain saturated fatty acid that inhibits the double-stranded DNA (dsDNA) binding activity of p53 DNA binding domain, with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans. It can be used as a tracer and as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-66999