Learn More
Medchemexpress LLC Docosanoic acid-d4-1 (Behenic acid-d4-1) | 1219804-98-4 | 99.0% | 344.61 | 5 MG
SDP
Catalog No. 5000454096
Encompass_Preferred
Docosanoic acid-d4-1 (Behenic acid-d4-1) is the deuterium-labeled Docosanoic acid. It is a long-chain saturated fatty acid that inhibits the double-stranded DNA (dsDNA) binding activity of the p53 DNA binding domain with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54096 Supplier Medchemexpress LLC Supplier No. HYW013049S45MG
Add to Cart
Add to Cart
Docosanoic acid-d4-1 (Behenic acid-d4-1) is the deuterium-labeled Docosanoic acid. It is a long-chain saturated fatty acid that inhibits the double-stranded DNA (dsDNA) binding activity of the p53 DNA binding domain with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.