Learn More
Medchemexpress LLC Docosanoic acid-d43 (Behenic acid-d43) | 29823-26-5 | 99.3% | 383.85 | 10 MG
SDP
Catalog No. 5000451233
Encompass_Preferred
Docosanoic acid-d43 (Behenic acid-d43) is the deuterium-labeled Docosanoic acid. Docosanoic acid is a long-chain saturated fatty acid. It inhibits the double-stranded DNA (dsDNA) binding activity of p53 DNA binding domain, with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51233 Supplier Medchemexpress LLC Supplier No. HYW013049S110MG
Docosanoic acid-d43 (Behenic acid-d43) is the deuterium-labeled Docosanoic acid. Docosanoic acid is a long-chain saturated fatty acid. It inhibits the double-stranded DNA (dsDNA) binding activity of p53 DNA binding domain, with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.