Learn More
Apexbio Technology LLC Docosanol (Abreua) 661-19-8 10g
SDP
Catalog No. 5000093857
Encompass_Preferred
Docosanol Abreua (CAS 661-19-8) is a small-molecule inhibitor targeting herpes simplex virus (HSV) It is designed to inhibit viral entry processes thereby preventing HSV replication at the initial stage of infection Docosanol Abreua exerts its biological activity primarily through the inhibition of viral envelope fusion with host cell membranes In cell-based studies Docosanol Abreua demonstrates antiviral inhibition with an IC50 value typically in the range of approximately 2 5-5 M Based on these pharmacological properties Docosanol Abreua holds research potential in antiviral mechanism studies investigation of viral entry pathways and host pathogen interaction assays
Catalog No. 50-000-93857 Supplier Apexbio Technology LLC Supplier No. A841610000
Docosanol Abreua (CAS 661-19-8) is a small-molecule inhibitor targeting herpes simplex virus (HSV) It is designed to inhibit viral entry processes thereby preventing HSV replication at the initial stage of infection Docosanol Abreua exerts its biological activity primarily through the inhibition of viral envelope fusion with host cell membranes In cell-based studies Docosanol Abreua demonstrates antiviral inhibition with an IC50 value typically in the range of approximately 2 5-5 M Based on these pharmacological properties Docosanol Abreua holds research potential in antiviral mechanism studies investigation of viral entry pathways and host pathogen interaction assays