Learn More
Cayman Chemical DocosapentaeNIc ACD mthyl 10mg
SDP
Catalog No. 502894440
Encompass_Preferred
An esterified form of docosapentaenoic acid; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils
Catalog No. 50-289-4440 Supplier Cayman Chemical Supplier No. 900187010 MG
An esterified form of docosapentaenoic acid; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils