Learn More
Cayman Chemical Docosapentaenoic Acid, Purity: ≥98%, CAS: 24880-45-3, 100 mg
SDP
Catalog No. 502895634
Encompass_Preferred
Docosapentaenoic acid is an ω-3 fatty acid found in fish oils. It is a minor constituent of the total serum unsaturated fatty acids in humans, ranging from 0.1 to 1%, and increases on dietary supplementation.{1936}
Catalog No. 50-289-5634 Supplier Cayman Chemical Supplier No. 90165100 MG
Docosapentaenoic acid is an ω-3 fatty acid found in fish oils. It is a minor constituent of the total serum unsaturated fatty acids in humans, ranging from 0.1 to 1%, and increases on dietary supplementation.{1936}