Learn More
Medchemexpress LLC HY-113159 5mg Medchemexpress, Docosapentaenoic acid (22n-3) CAS:24880-45-3 Purity:>98%
SDP
Catalog No. 502080842
Encompass_Preferred
Medchemexpress, HY-113159 5mg Docosapentaenoic acid (22n-3) CAS:24880-45-3 Docosapentaenoic acid (22n-3) is a component of phospholipids found in all animal cell membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0842 Supplier Medchemexpress LLC Supplier No. HY1131595MG
Medchemexpress, HY-113159 5mg Docosapentaenoic acid (22n-3) CAS:24880-45-3 Docosapentaenoic acid (22n-3) is a component of phospholipids found in all animal cell membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.