Learn More
Cayman Chemical DocosapentaenoIc AcId-d5 100ug
SDP
Catalog No. 502983266
Encompass_Preferred
An internal standard for the quantification of DPA by GC- or LC-MS
Catalog No. 50-298-3266 Supplier Cayman Chemical Supplier No. 20913100 UG
An internal standard for the quantification of DPA by GC- or LC-MS