Learn More
Cayman Chemical Docosatrienoic Acid methyl ester, Purity: ≥98%, CAS: 108698-01-7, 10 mg
SDP
Catalog No. 502982926
Encompass_Preferred
An ester version of docosatrienoic acid, a rare ω-3 fatty acid that inhibits [3H]-LTB4 binding to pig neutrophils at a concentration of 5 µM
Catalog No. 50-298-2926 Supplier Cayman Chemical Supplier No. 2056710 MG
An ester version of docosatrienoic acid, a rare ω-3 fatty acid that inhibits [3H]-LTB4 binding to pig neutrophils at a concentration of 5 µM