Learn More
Cayman Chemical Docosatrienoic Acid, Purity: ≥98%, CAS: 28845-86-5, 100 mg
SDP
Catalog No. 502895635
Encompass_Preferred
A rare ω-3 fatty acid; not readily detected in normal phospholipid PUFA pools; polyunsaturated version of docosanoic acid; inhibits [3H]-LTB4 binding to pig neutrophils at a concentration of 5 µM; inhibits mammalian DNA polymerases and human topoisomerases I and II
Catalog No. 50-289-5635 Supplier Cayman Chemical Supplier No. 90170100 MG
A rare ω-3 fatty acid; not readily detected in normal phospholipid PUFA pools; polyunsaturated version of docosanoic acid; inhibits [3H]-LTB4 binding to pig neutrophils at a concentration of 5 µM; inhibits mammalian DNA polymerases and human topoisomerases I and II