Learn More
Cayman Chemical Docosatetraenoyl Ethanolamide, Purity: ≥98%, CAS: 150314-35-5, 50 mg
SDP
Catalog No. 502894862
Encompass_Preferred
An endocannabinoid containing docosatetraenoic acid in place of the arachidonate moiety of AEA; acts on CB1 receptors with a potency and efficacy similar to that of AEA but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated
Catalog No. 50-289-4862 Supplier Cayman Chemical Supplier No. 9038550 MG
An endocannabinoid containing docosatetraenoic acid in place of the arachidonate moiety of AEA; acts on CB1 receptors with a potency and efficacy similar to that of AEA but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated