Learn More
Cayman Chemical Docosanoic Acid, Purity: ≥95%, CAS: 112-85-6, 500 mg
SDP
Catalog No. 502896867
Encompass_Preferred
A long-chain saturated fatty acid; binds to the DNA binding domain of p53 (Kd = 12 nM) and inhibits the DNA binding activity; inhibits rat DNA polymerase β and human DNA polymerase λ activity at 100 µM; inhibits human DNA topoisomerase I and II relaxation activity at 25 µM; liver levels are reduced in rats fed a high-fat or a high-fat high-cholesterol diet
Catalog No. 50-289-6867 Supplier Cayman Chemical Supplier No. 9000338500 MG
A long-chain saturated fatty acid; binds to the DNA binding domain of p53 (Kd = 12 nM) and inhibits the DNA binding activity; inhibits rat DNA polymerase β and human DNA polymerase λ activity at 100 µM; inhibits human DNA topoisomerase I and II relaxation activity at 25 µM; liver levels are reduced in rats fed a high-fat or a high-fat high-cholesterol diet