Learn More
Cayman Chemical Docosanoic Acid-d43, Purity: ≥99% deuterated forms (d1-d43), CAS: 29823-26-5, 1 mg
SDP
Catalog No. 502940569
Encompass_Preferred
An internal standard for the quantification of docosanoic acid by GC- or LC-MS
Catalog No. 50-294-0569 Supplier Cayman Chemical Supplier No. 290751 MG
An internal standard for the quantification of docosanoic acid by GC- or LC-MS