Learn More
Cayman Chemical DocosnoIc AcId-d43 5mg
SDP
Catalog No. 502941343
Encompass_Preferred
An internal standard for the quantification of docosanoic acid by GC- or LC-MS
Catalog No. 50-294-1343 Supplier Cayman Chemical Supplier No. 290755 MG
An internal standard for the quantification of docosanoic acid by GC- or LC-MS