Learn More
Cayman Chemical Docosanoic Acid methyl ester, Purity: ≥95%, CAS: 929-77-1, 1 g
SDP
Catalog No. 502933779
Encompass_Preferred
An ester form of docosanoic acid; has been found in soybean, corn, sunflower, and canola oils, as well as beef fat, used in the production of biodiesel
Catalog No. 50-293-3779 Supplier Cayman Chemical Supplier No. 268661 G
An ester form of docosanoic acid; has been found in soybean, corn, sunflower, and canola oils, as well as beef fat, used in the production of biodiesel